missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl sulfone, 98%

Product Code. 11420367
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11420367 250 g 250g
11410367 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11420367 Supplier Thermo Scientific Alfa Aesar Supplier No. B21747.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 67-71-0 | C2H6O2S | 94.13 g/mol

Dimethyl sulfone is used as a high-temperature solvent for both inorganic and organic substances. It acts as an intermediate as well as medium in organic synthesis. It plays an important role as a carbon source by the airborne bacteria Afipia. It is also employed as an extraction solvent for the separation of aromatic compounds like benzene, toluene and xylenes from aliphatic hydrocarbons. Further, it is used in the polymerization process. It finds application in pharmaceuticals, agrochemicals, paint and coating materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl sulfone is used as a high-temperature solvent for both inorganic and organic substances. It acts as an intermediate as well as medium in organic synthesis. It plays an important role as a carbon source by the airborne bacteria Afipia. It is also employed as an extraction solvent for the separation of aromatic compounds like benzene, toluene and xylenes from aliphatic hydrocarbons. Further, it is used in the polymerization process. It finds application in pharmaceuticals, agrochemicals, paint and coating materials.

Solubility
Soluble in water, methanol, ethanol and acetone.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-71-0
Molecular Formula C2H6O2S
Molecular Weight (g/mol) 94.13
MDL Number MFCD00007566
InChI Key HHVIBTZHLRERCL-UHFFFAOYSA-N
Synonym dimethyl sulfone, methyl sulfone, sulfonylbismethane, dimethylsulfone, dimethyl sulphone, methane, sulfonylbis, sulphonylbismethane, methylsulfone, methanesulfonylmethane, sulfonyldimethane
PubChem CID 6213
ChEBI CHEBI:9349
IUPAC Name methylsulfonylmethane
SMILES CS(C)(=O)=O

Specifications

Melting Point 108°C to 111°C
Density 1.45
Boiling Point 238°C
Flash Point 143°C (289°F)
Odor Odorless
Quantity 250 g
Beilstein 1737717
Merck Index 14,3258
Solubility Information Soluble in water,methanol,ethanol and acetone.
Formula Weight 94.13
Percent Purity 99%
Chemical Name or Material Dimethyl sulfone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.