missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Sulfone, TRC

Product Code. 30372372
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30372372 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30372372 Supplier Toronto Research Chemicals Supplier No. TRCD47935010G
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl Sulfone (MSM)
CAS 67-71-0
Molecular Formula C2 H6 O2 S
Quantity 10 g
Synonym Dimethyl Sulfone, Methane, 1,1'-sulfonylbis-, Methane, sulfonylbis- (9CI), Methyl sulfone (6CI,8CI), 1,1'-Sulfonylbis[methane], Dimethyl sulfone, Dimethyl sulphone, Lignisul MSM, MSM, Methylsulfonylmethane, NSC 63345, Opti MSM, Opti-dimethyl sulfone
InChI Formula InChI=1S/C2H6O2S/c1-5(2,3)4/h1-2H3
SMILES CS(=O)(=O)C
IUPAC Name methylsulfonylmethane
Molecular Weight (g/mol) 94.13
Formula Weight 94.0089
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.