missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Sulfoxide (99+% Purity), MP Biomedicals™

Product Code. 11401611 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11401611 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11401611 Supplier MP Biomedicals Supplier No. 0219018691

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dimethyl Sulfoxide is used to enhance dermal absorption of many chemicals.

  • Dimethyl sulfoxide is used to enhance dermal absorption of many chemicals.
  • A solvent for many organic and inorganic compounds including fats, carbohydrates, dyes, resins, and polymers.
  • Used in antifreeze or hydraulic fluids and as a cryopreservative for cell cultures.
  • Used in the oxidation of thiols and disulfides to sulfonic acids.
  • Used as a PCR cosolvent to help improve yields, especially in long PCR.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-68-5
Color Colorless
Density 1.104 g/cm3 (at 20°C)
Quantity 1 L
Physical Form Liquid To Solid
Assay Percent Range >99%
Molecular Formula C2H6OS
MDL Number MFCD00002089
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
SMILES CS(C)=O
IUPAC Name methylsulfinylmethane
Molecular Weight (g/mol) 78.13
PubChem CID 679
ChEBI CHEBI:28262
Formula Weight 78.13
Chemical Name or Material Dimethyl sulfoxide
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.