missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Sulfoxide, Certified AR for Analysis, Fisher Chemical™

Product Code. 11403893 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
10 L
25 L
Packaging:
HDPE Bottle
Plastic Container
Unit Size:
1 L
10 L
2.5 L
25 L
5 L
500mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11403893 5 L HDPE Bottle 5 L
10213810 500 mL HDPE Bottle 500mL
10206581 1 L HDPE Bottle 1 L
10162090 2.5 L HDPE Bottle 2.5 L
10194423 10 L Plastic Container 10 L
10318153 25 L Plastic Container 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 11403893 Supplier Fisher Chemical Supplier No. D/4121/21
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

C2H6OS, CAS Number-67-68-5, demasorb, demsodrox, methyl sulfoxide, dimexide, dimethyl sulfoxide, dimethylsulfoxide, demavet, methane, sulfinylbis, dimethyl sulphoxide, dmso, 2.5L, 189 deg.C, CHEBI:28262, Colorless, 78.13g/mol, IAZDPXIOMUYVGZ-UHFFFAOYSA-N, methylsulfinylmethane, 18.4 deg.C, 2089 | CAS: 67-68-5 | C2H6OS | 78.13 g/mol

C2H6OS, CAS Number-67-68-5, demasorb, demsodrox, methyl sulfoxide, dimexide, dimethyl sulfoxide, dimethylsulfoxide, demavet, methane, sulfinylbis, dimethyl sulphoxide, dmso, 2.5L, 189 deg.C, CHEBI:28262, Colorless, 78.13g/mol, IAZDPXIOMUYVGZ-UHFFFAOYSA-N, methylsulfinylmethane, 18.4 deg.C, 2089

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
SMILES CS(C)=O

Specifications

Melting Point 18.4°C
Color Colorless
Quantity 5 L
Packaging HDPE Bottle
Physical Form Liquid
Boiling Point 189°C
IUPAC Name methylsulfinylmethane
Formula Weight 78.13g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.