missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl sulfoxide, CHROMASOLV™ Plus, for HPLC, ≥99.7%, Solstice

Product Code. 15624630
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
2 L
2.5 L
4 x 2.5 L
4 x 4 L
6 x 1 L
Unit Size:
1 L
100mL
2 L
2.5 L
500mL
Pack of 4
Pack of 6
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15624630 1 L 1 L
15604630 100 mL 100mL
15674630 500 mL 500mL
15644630 2 L 2 L
15634630 2.5 L 2.5 L
15684630 6 x 1 L Pack of 6
15654630 4 x 2.5 L Pack of 4
15664630 4 x 4 L Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Supercools easily and remelts slowly at room temperature. Solidified product can be re-liquified by warming to room temperature without detriment to the product.
Product Code. 15624630 Supplier Honeywell-Chromasolv Supplier No. 348691L
Solstice Chromasolv Promotion
Use promo code 27944
View all promotions
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Supercools easily and remelts slowly at room temperature. Solidified product can be re-liquified by warming to room temperature without detriment to the product.

CHROMASOLV™ Plus, for HPLC,≥99.7%

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
SMILES CS(C)=O

Specifications

Melting Point 16°C to 19°C
Density 1.10g/mL
Quantity 1 L
Refractive Index n20/D 1.479
Linear Formula (CH3)2SO
Vapor Pressure 0.42mmHg (20°C)
Beilstein 506008
Boiling Point 189°C
IUPAC Name methylsulfinylmethane
Formula Weight 78.13g/mol
Percent Purity ≥99.7%
Grade CHROMASOLV™ Plus
Chemical Name or Material Dimethyl sulfoxide, for HPLC
Show More Show Less
Product Identifier
  • Dimethyl sulfoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.