missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Sulfoxide, Extra Pure, SLR, Fisher Chemical™

Product Code. 10589950 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 mL
2.5 L
5 L
25 L
Packaging:
HDPE Bottle
Plastic Container
Unit Size:
2.5 L
25 L
5 L
500mL
This item is not returnable. View return policy

Product Code. 10589950

Brand: Fisher Chemical D/4120/25

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

C2H6OS, CAS Number-67-68-5, demsodrox, methyl sulfoxide, dimexide, methane, sulfinylbis, dimethyl sulfoxide, dmso, demavet, dimethylsulfoxide, dimethyl sulphoxide, demasorb, 500mL, 189 deg.C, CHEBI:28262, Colorless, 78.13g/mol, IAZDPXIOMUYVGZ-UHFFFAOYSA-N, methylsulfinylmethane, 18.4 deg.C, 2089

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
IUPAC Name methylsulfinylmethane
SMILES CS(C)=O

Specifications

Melting Point 18.4°C
Color Colorless
Boiling Point 189°C
Quantity 25 L
Packaging Plastic Container
Physical Form Liquid
Vapor Pressure 0.55mbar at 20°C
Viscosity 1.98 mPaS at 25°C
Formula Weight 78.13g/mol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.