missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl sulfoxide, Reagent Grade, Honeywell

Product Code. 15637830 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
1 gA
4 L
5 L
25 L
230 kg
Unit Size:
1 GAL
1 L
100mL
2.5 L
230kg
25 L
4 L
5 L
500mL
Product Code. Quantity unitSize
15637830 500 mL 500mL
15667820 100 mL 100mL
15697820 1 L 1 L
15607830 2.5 L 2.5 L
15698480 4 L 4 L
15647830 5 L 5 L
15627830 25 L 25 L
15687820 1 gA 1 GAL
15617830 230 kg 230kg
9 options
This item is not returnable. View return policy

Product Code. 15637830

Brand: Honeywell Research Chemicals D5879500ML

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
SMILES CS(C)=O

Specifications

Density 1.10g/mL
Quantity 500 mL
Explosion Limit 42%, 63°F
Flash Point 87°C (188.6°F)
Ignition Point 573°F
Linear Formula (CH3)2SO
Vapor Pressure 0.42 mmHg (20°C)
Beilstein 506008
IUPAC Name methylsulfinylmethane
Formula Weight 78.13g/mol
Percent Purity ≥99.5%
Grade Reagent
Chemical Name or Material Dimethyl sulfoxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.