missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl sulfoxide, Honeywell Riedel-de Haën™

Product Code. 15665297 Shop All Honeywell Research Chemicals Products
Click to view available options
:
25 L
This item is not returnable. View return policy

Product Code. 15665297

Brand: Honeywell Research Chemicals 4164025L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, ≥99.9% (GC)

ACS General Use Solvents

  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Specifications

pH Not applicable
Melting Point 18.5°C
CAS 67-68-5
Color Colorless
Boiling Point 189°C
Quantity 25 L
Packaging Plastic Pail
Molecular Formula C2H6OS
Synonym DMSO
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
SMILES CS(C)=O
IUPAC Name methanesulfinylmethane
Molecular Weight (g/mol) 78.13
Percent Purity 0.999
Grade ACS
Chemical Name or Material Dimethyl sulfoxide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.