missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl sulfoxide, Spectrophotometric Grade, 99.9+%

Product Code. 11368357
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
Unit Size:
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368357 1 L 1 L
11358357 4 x 1 L 4 L
11378357 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11368357 Supplier Thermo Scientific Alfa Aesar Supplier No. 032434.K2
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 67-68-5 | C2H6OS | 78.13 g/mol

Dimethyl sulfoxide is used in spectrophotometry and environmental testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
SMILES CS(C)=O

Specifications

Melting Point 18.4°C
Color Colorless
Appearance Colorless liquid
Density 1.101 g/cm3 at 20°C
Quantity 1 L
Flash Point 87°C (189°F)
Odor Mild
Physical Form Liquid
Refractive Index 1.479
Assay Percent Range >99.9%
Vapor Pressure 2.5 hPa (2mm Hg) at 20°C
Beilstein 506008
Boiling Point 189°C
Solubility Information Miscible with water,ethanol,ether,benzene,chloroform and acetone.
IUPAC Name methylsulfinylmethane
Formula Weight 78.13
Grade Spectrophotometry
Chemical Name or Material Dimethyl sulfoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.