missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Sulfoxide, TRC

Product Code. 30378977
Change view
Click to view available options
Quantity:
5 mL
100 mL
250 mL
Unit Size:
100mL
250mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30378977 100 mL 100mL
30374603 5 mL 5mL
30372966 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30378977 Supplier Toronto Research Chemicals Supplier No. TRCD479380100ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dimethylsulfoxide
CAS 67-68-5
Molecular Formula C2 H6 O S
Quantity 100 mL
Synonym Dimethyl sulfoxide, Sulfinylbismethane, Methylsulfinylmethane, Methane, sulfinylbis- (9CI), Methyl sulfoxide (8CI), DMS 70, DMS 90, DMSO, DMSO Evol, Demasorb, Demavet, Demeso, Demsodrox, Dimethyl sulfoxide, Dimethyl sulphoxide, Dimethylsulfoxide, Dimexide, Dimexidum, Dipirartril-tropico, Dolicur, Domoso, Dromisol, Durasorb, Gamasol 90, Herpid, Hyadur, Infiltrina, Kemsol, NSC 763, Rimso 50, SQ 9453, Sclerosol, Somipront, Sulfinylbismethane, Syntexan, Methane, 1,1'-sulfinylbis-
InChI Formula InChI=1S/C2H6OS/c1-4(2)3/h1-2H3
SMILES CS(=O)C
IUPAC Name methylsulfinylmethane
Molecular Weight (g/mol) 78.13
Formula Weight 78.0139
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.