missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl sulphoxide, BAKER ANALYZED™, J.T. Baker™

Product Code. 15568654 Shop All J.T.Baker Products
Change view
Click to view available options
:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15568654 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15568654 Supplier JT Baker Supplier No. 9754.1000

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For headspace gas chromatography

Specifications

CAS 67-68-5
Acidity max. 0.0005 meq/g
Quantity 1 L
Assay Percent Range min. 99.9%
Residue after Evaporation max. 0.0005%
Molecular Formula C2H6OS
MDL Number MFCD00002089
Water max. 0.05%
Synonym DMSO , Methyl sulphoxide
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
SMILES CS(C)=O
IUPAC Name methanesulfinylmethane
Molecular Weight (g/mol) 78.13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.