missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl trisulfide, 98%

Product Code. 15455687
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15455687 25 g 25g
15445687 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15455687 Supplier Thermo Scientific Alfa Aesar Supplier No. H32913.14
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 3658-80-8 | C2H6S3 | 126.25 g/mol

Dimethyl trisulfide is reported as a major aroma component in cooked Brassicaceous vegetables. Trap baits containing dimethyl trisulfide have been used to capture Calliphora loewi and other blowflies. Dimethyl trisulfide along with dimethyl sulfide and dimethyl disulfide have been confirmed as volatile compounds given off by the fly-attracting plant known as dead-horse arum (Helicodiceros muscivorus). These flies are attracted to the odor of fetid meat and help pollinate this plant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl trisulfide is reported as a major aroma component in cooked Brassicaceous vegetables. Trap baits containing dimethyl trisulfide have been used to capture Calliphora loewi and other blowflies. Dimethyl trisulfide along with dimethyl sulfide and dimethyl disulfide have been confirmed as volatile compounds given off by the fly-attracting plant known as dead-horse arum (Helicodiceros muscivorus). These flies are attracted to the odor of fetid meat and help pollinate this plant.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at 4°C. Store in an air-tight and light-resistant container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3658-80-8
Molecular Formula C2H6S3
Molecular Weight (g/mol) 126.25
MDL Number MFCD00039808
InChI Key YWHLKYXPLRWGSE-UHFFFAOYSA-N
Synonym dimethyl trisulfide, trisulfide, dimethyl, methyl trisulfide, 2,3,4-trithiapentane, dimethyl trisulphide, dimethyltrisulfide, methyltrisulfanyl methane, dimethyltrisulfane, unii-3e691t3nl1, dmts
PubChem CID 19310
ChEBI CHEBI:4614
IUPAC Name (methyltrisulfanyl)methane
SMILES CSSSC

Specifications

Density 1.203
Boiling Point 165°C to 175°C
Flash Point 52°C (126°F)
Odor Stench
Linear Formula CH3SSSCH3
Refractive Index 1.6
Quantity 25 g
UN Number UN1993
Solubility Information Insoluble in water.
Formula Weight 126.26
Percent Purity 98%
Chemical Name or Material Dimethyl trisulfide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dimethyl trisulfide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Dimethyl trisulfide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.