missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Yellow Indicator, Solstice

Product Code. 15633910 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15633910 25 g Glass Bottle 25g
15623910 100 g Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15633910 Supplier Honeywell-Fluka Supplier No. 3312925G
  • ONLINE EXCLUSIVE PRICE
    86.00 €
    72.30 € / 25g
    Save 13.70 €
    15% Off
 Restricted Item
Estimated Shipment: 24-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Indicator

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-11-7
Molecular Formula C14H15N3
Molecular Weight (g/mol) 225.30
MDL Number MFCD00008308
InChI Key JCYPECIVGRXBMO-UHFFFAOYSA-N
PubChem CID 6053
ChEBI CHEBI:17903
IUPAC Name N,N-dimethyl-4-phenyldiazenylaniline
SMILES CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1

Specifications

Melting Point 111°C
Odor Odourless
Packaging Glass Bottle
Linear Formula C6H5N=NC6H4N(CH3)2
UN Number UN2811
Beilstein 746016
Color Yellow
Solubility Information Insoluble in water
pH 5 to 7 (at 10.00 g/L 20°C)
Quantity 25 g
Formula Weight 225.29g/mol
Grade Indicator
Chemical Name or Material Dimethyl Yellow
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dimethyl yellow
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 4-Dimethylaminoazobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.