missing translation for 'onlineSavingsMsg'
Learn More

Dimethylamine, 26 wt.% solution in water, Thermo Scientific Chemicals

Product Code. 10166890 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10166890 1 kg 1kg
15274724 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10166890 Supplier Thermo Scientific Acros Supplier No. 408000010
  • ONLINE EXCLUSIVE PRICE
    40.40 €
    37.70 € / 1kg
    Save 2.70 €
    6% Off
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dimethylamine
CAS 124-40-3, 7732-18-5
Melting Point -37.0°C
Color Colorless
Density 0.8900g/mL
Boiling Point 54.0°C
Flash Point −18°C
Packaging Glass bottle
Molecular Formula C2H7N
Linear Formula (CH3)2NH
Refractive Index 1.37
MDL Number MFCD00008288
Quantity 1 kg
Beilstein 04,39
Fieser 07,119
Merck Index 15,325
Specific Gravity 0.89
Synonym dimethylamine, n,n-dimethylamine, methanamine, n-methyl, dimethylamin, dimethyl amine, dimethyl-amine, dimethylamine anhydrous, rcra waste number u092, dimethylamine solution
Solubility Information Solubility in water: >500g/L (20°C). Other solubilities: soluble in alcohol and ether
InChI Key ROSDSFDQCJNGOL-UHFFFAOYSA-N
SMILES CNC
IUPAC Name N-methylmethanamine
Molecular Weight (g/mol) 45.07
PubChem CID 674
ChEBI CHEBI:17170
Formula Weight 45.07
Percent Purity 25 to 27%
Physical Form Liquid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.