missing translation for 'onlineSavingsMsg'
Learn More

Dimethylamine hydrochloride, 99%, Thermo Scientific Chemicals

Product Code. 10102970 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
500 g
1 kg
2.5 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
1kg
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10102970 1 kg Plastic bottle 1kg
10041790 5 g Glass bottle 5g
10122970 500 g Plastic bottle 500g
10226883 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10102970 Supplier Thermo Scientific Acros Supplier No. 155270010
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 506-59-2
MDL Number MFCD00012477
InChI Key IQDGSYLLQPDQDV-UHFFFAOYSA-N
Synonym dimethylamine hydrochloride, n-methylmethanamine hydrochloride, dimethylammonium chloride, methanamine, n-methyl-, hydrochloride, dimethylamine hcl, dimethylaminehydrochloride, unii-7m4cwb6aok, dimethylamine, hydrochloride, hydrochloric acid dimethylamine, n,n-dimethylamine hydrochloride
PubChem CID 10473

Specifications

Melting Point 160.0°C
Color White
pH 5 to 6 (5% aq. soln.)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (on dry substance) (Argentometry)
Packaging Plastic bottle
Molecular Formula C2H7N·HCl
Linear Formula (CH3)2NH·HCl
Quantity 1 kg
Beilstein 04, 39
Merck Index 15, 3250
Solubility Information Solubility in water: 3000g/L (20°C). Other solubilities: soluble in alcohol,chloroform,practically insoluble in ether
SMILES CNC.Cl
IUPAC Name N-methylmethanamine;hydrochloride
Molecular Weight (g/mol) 81.55
Formula Weight 81.55
Percent Purity 99%
Insoluble Matter 0.005% max. (in water)
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Dimethylamine hydrochloride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Dimethylamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.