missing translation for 'onlineSavingsMsg'
Learn More

Dimethylamine solution purum 33% in absolute ethanol (~5.6 M), TRC

Product Code. 30369034
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30369034 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30369034 Supplier Toronto Research Chemicals Supplier No. TRCD461481100ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dimethylamine
CAS 124-40-3
Molecular Formula C2H7N
Quantity 100 mL
Synonym Dimethylamine, N-Methylmethanamine, N,N-Dimethylamine, NSC 8650
InChI Formula InChI=1S/C2H7N/c1-3-2/h3H,1-2H3
SMILES CNC
Molecular Weight (g/mol) 45.08
Concentration or Composition (by Analyte or Components) 33 %
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.