missing translation for 'onlineSavingsMsg'
Learn More

Dimethylcyanamide, 97%, Thermo Scientific Chemicals

Product Code. 10393902 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10393902 5 mL Glass bottle 5mL
10308150 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10393902 Supplier Thermo Scientific Acros Supplier No. 200210050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1467-79-4
Molecular Formula C3H6N2
Molecular Weight (g/mol) 70.10
MDL Number MFCD00001767
InChI Key OAGOUCJGXNLJNL-UHFFFAOYSA-N
Synonym dimethyl cyanamide, cyanamide, dimethyl, n-cyano-n-methylmethanamine, dimethylkyanamid, cyanodimethylamine, n-cyanodimethylamine, dimethylkyanamid czech, unii-6p25ipq8gn, ccris 5909, cyanamide, n,n-dimethyl
PubChem CID 15112
IUPAC Name dimethylcyanamide
SMILES CN(C)C#N

Specifications

Melting Point -41.0°C
Color Colorless to Yellow
Density 0.8670g/mL
Boiling Point 161.0°C to 163.0°C
Flash Point 58°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (CH3)2NCN
Refractive Index 1.4090 to 1.4110
Quantity 5 mL
Beilstein 04, 74
Specific Gravity 0.867
Solubility Information Solubility in water: miscible. Other solubilities: soluble in acetone,ethyl ether,ethnaol
Formula Weight 70.09
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Dimethylcyanamide, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dimethylcyanamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Flammable liquid Category 3
Hazard Statement
  • H310-Fatal in contact with skin.
  • H301-Toxic if swallowed.
  • H331-Toxic if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.