missing translation for 'onlineSavingsMsg'
Learn More

Dimethyldichlorosilane, Pure, Fisher Chemical™

Product Code. 10636642 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10636642 250 mL 250mL
10498590 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10636642 Supplier Fisher Chemical Supplier No. D/3820/PB07
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 75-78-5
Molecular Formula C2H6Cl2Si
Molecular Weight (g/mol) 129.055
MDL Number 491
InChI Key LIKFHECYJZWXFJ-UHFFFAOYSA-N
Synonym dimethyldichlorosilane, silane, dichlorodimethyl, dichloro dimethyl silane, inerton aw-dmcs, dichlorodimethylsilicon, dimethyl-dichlorsilan, inerton dw-dmc, repel-silan, dimethylsilane dichloride, dimethyl dichlorosilane
PubChem CID 6398
IUPAC Name dichloro(dimethyl)silane
SMILES C[Si](C)(Cl)Cl

Specifications

Melting Point -76°C
Boiling Point 70°C
Packaging Plastic Coated Glass bottle
Quantity 250 mL
Formula Weight 129.06g/mol
Vapor Pressure 150 hPa at 20°C
Physical Form Liquid
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dichlorodimethylsilane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.