missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Dimethylenastron
GREENER_CHOICE

Product Code. 18750268
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18750268 50 mg 50mg
18773168 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18750268 Supplier Tocris Bioscience™ Supplier No. 5261/50
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Inhibitor of mitotic motor kinesin Eg5

Dimethylenastron is an inhibitor of mitotic motor kinesin Eg5 (IC50 = 200 nM). Arrests mitosis, triggers apoptosis and up-regulates Hsp-70 expression in multiple human myeloma cell lines. Also inhibits the growth of several types of cancer cell lines (EC50 values are 330, 603, 743 and 881 nM in HCT116, Htert-HME1, BxPC3, K562 and NCI-H1299 cancer cells, respectively). Suppresses the migration and invasion of PANC1 cells in vitro. Cytoprotective.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,3,4,6,7,8-Hexahydro-4-(3-hydroxyphenyl)-7,7-dimethyl-2-thioxo-5(1H)-quinazolinone
CAS 863774-58-7
Quantity 50 mg
Target Kinesin Inhibitors
Molecular Formula C16H18N2O2S
Formula Weight 302.39
Purity >99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.