Learn More
Tocris Bioscience™ Dimethylenastron
GREENER_CHOICE
Inhibitor of mitotic motor kinesin Eg5
Brand:  Tocris Bioscience™ 5261/10
Encompass
Product Code. 15776626
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Dimethylenastron is an inhibitor of mitotic motor kinesin Eg5 (IC50 = 200 nM). Arrests mitosis, triggers apoptosis and up-regulates Hsp-70 expression in multiple human myeloma cell lines. Also inhibits the growth of several types of cancer cell lines (EC50 values are 330, 603, 743 and 881 nM in HCT116, Htert-HME1, BxPC3, K562 and NCI-H1299 cancer cells, respectively). Suppresses the migration and invasion of PANC1 cells in vitro. Cytoprotective.
Specifications
Specifications
2,3,4,6,7,8-Hexahydro-4-(3-hydroxyphenyl)-7,7-dimethyl-2-thioxo-5(1H)-quinazolinone
863774-58-7