missing translation for 'onlineSavingsMsg'
Learn More

Dimethylethylhexadecylammonium bromide, 99%

Product Code. 10532041 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
This item is not returnable. View return policy

Product Code. 10532041

Brand: Thermo Scientific Acros 211871000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 124-03-8 | C20H44BrN | 378.483 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-03-8
Molecular Formula C20H44BrN
Molecular Weight (g/mol) 378.483
MDL Number MFCD00011774
InChI Key VUFOSBDICLTFMS-UHFFFAOYSA-M
Synonym Cetyldimethylethylammonium bromide
PubChem CID 31280
IUPAC Name ethyl-hexadecyl-dimethylazanium;bromide
SMILES CCCCCCCCCCCCCCCC[N+](C)(C)CC.[Br-]

Specifications

Color White
Physical Form Powder
pH 6 to 8 (1% soln.)
Quantity 100 g
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 99%
Linear Formula CH3(CH2)15N(CH3)2(C2H5)Br
Beilstein 04, III, 425
Merck Index 15, 2028
Formula Weight 378.48
Percent Purity ≥98.5%
Chemical Name or Material Dimethylethylhexadecylammonium bromide, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Dimethylethylhexadecylammonium bromide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.