missing translation for 'onlineSavingsMsg'
Learn More

Dimethylgermanium dichloride, 97+%

Product Code. 11377778
Change view
Click to view available options
Quantity:
0.5 g
2 g
10 g
Unit Size:
0.5g
10g
2g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377778 0.5 g 0.5g
11357778 2 g 2g
11367778 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11377778 Supplier Thermo Scientific Alfa Aesar Supplier No. 034107.02
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1529-48-2 | C2H6Cl2Ge | 173.6 g/mol

Dimethylgermanium dichloride is a useful organogermanium compound for synthesis and proteomics research. It is also used as an organic intermediate and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1529-48-2
Molecular Formula C2H6Cl2Ge
Molecular Weight (g/mol) 173.6
MDL Number MFCD00013588
InChI Key YQECBLVSMFAWIZ-UHFFFAOYSA-N
Synonym dimethylgermanium dichloride, germane, dichlorodimethyl, dimethyldichlorogermane, dichloro dimethyl germane, dimethylgermanedichloride, dimethylgermyl dichloride, geme2cl2, acmc-1boyh, germanium dimethyl dichloride, dimethyl germanium dichloride
PubChem CID 73724
IUPAC Name dichloro(dimethyl)germane
SMILES C[Ge](C)(Cl)Cl

Specifications

Melting Point -22°C
Density 1.51 g/mL
Boiling Point 124°C
Flash Point 32°C (90°F)
Refractive Index 1.46
Quantity 0.5 g
UN Number UN2924
Sensitivity Moisture sensitive
Solubility Information Hydrolyses in water.
Formula Weight 173.61
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material Dimethylgermanium dichloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.