missing translation for 'onlineSavingsMsg'
Learn More

Dimethylglyoxime, 99+%, ACS reagent

Product Code. 10625491 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10625491 100 g Plastic bottle 100g
10615491 25 g Plastic bottle 25g
10015700 500 g Plastic bottle 500g
10751544 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10625491 Supplier Thermo Scientific Acros Supplier No. 408331000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 95-45-4 | C4H8N2O2 | 116.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 95-45-4
Molecular Formula C4H8N2O2
Molecular Weight (g/mol) 116.12
MDL Number MFCD00002117
InChI Key JGUQDUKBUKFFRO-GGWOSOGESA-N
Synonym biacetyl dioxime, dimethylglyoxime, diacetyldioxime, 2,3-butanedione dioxime, diacetyl dioxime, 2,3-diisonitrosobutane, biacetyl, dioxime, chugaev's reagent, nsc 9, unii-2971mft1ky
PubChem CID 5356010
SMILES C\C(=N/O)\C(\C)=N\O

Spécification

Melting Point 239.0°C to 242.0°C
Color White
Ignition Residue 0.05% max.
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic bottle
Linear Formula CH3C(=NOH)C(=NOH)CH3
Quantity 100 g
Beilstein 01, 772
Merck Index 15, 3270
Solubility Information Solubility in water: 0,6g/L 20°C. Other solubilities: soluble in alcohol,ether,pyridine and acetone,slightly soluble in benzene
Sublimation Point >70°C
IUPAC Name N-[(E)-3-nitrosobut-2-en-2-yl]hydroxylamine
Formula Weight 116.12
Percent Purity 99+%
Grade ACS Reagent
Insoluble Matter 0.02% max. (in ethanol)
Physical Form Crystalline Powder
Chemical Name or Material Dimethylglyoxime
Afficher plus Afficher moins
compliance-icons compliance-icons
Identifiant produit
  • Dimethylglyoxime
Mention d'avertissement
  • Danger
Catégorie de danger
  • Flammable solid Category 2
  • Acute toxicity Category 3
Mention de danger
  • H228-Flammable solid.
  • H301-Toxic if swallowed.
Conseils de prudence
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.