missing translation for 'onlineSavingsMsg'
Learn More

Dimethylglyoxime, 99+%, ACS reagent

Product Code. 10751544 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10751544 2.5 kg Plastic bottle 2.5kg
10615491 25 g Plastic bottle 25g
10625491 100 g Plastic bottle 100g
10015700 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10751544 Supplier Thermo Scientific Acros Supplier No. 408330025
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 95-45-4 | C4H8N2O2 | 116.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-45-4
Molecular Formula C4H8N2O2
Molecular Weight (g/mol) 116.12
MDL Number MFCD00002117
InChI Key JGUQDUKBUKFFRO-GGWOSOGESA-N
Synonym biacetyl dioxime, dimethylglyoxime, diacetyldioxime, 2,3-butanedione dioxime, diacetyl dioxime, 2,3-diisonitrosobutane, biacetyl, dioxime, chugaev's reagent, nsc 9, unii-2971mft1ky
PubChem CID 5356010
SMILES C\C(=N/O)\C(\C)=N\O

Specifications

Melting Point 239.0°C to 242.0°C
Color White
Ignition Residue 0.05% max.
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic bottle
Linear Formula CH3C(=NOH)C(=NOH)CH3
Quantity 2.5 kg
Beilstein 01, 772
Merck Index 15, 3270
Solubility Information Solubility in water: 0,6g/L 20°C. Other solubilities: soluble in alcohol,ether,pyridine and acetone,slightly soluble in benzene
Sublimation Point >70°C
IUPAC Name N-[(E)-3-nitrosobut-2-en-2-yl]hydroxylamine
Formula Weight 116.12
Percent Purity 99+%
Grade ACS Reagent
Insoluble Matter 0.02% max. (in ethanol)
Physical Form Crystalline Powder
Chemical Name or Material Dimethylglyoxime
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.