missing translation for 'onlineSavingsMsg'
Learn More

Dimethylglyoxime, 99+%

Product Code. 13418418 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13418418 2.5 kg Plastic bottle 2.5kg
10408150 100 g Plastic bottle 100g
10012170 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 13418418 Supplier Thermo Scientific Acros Supplier No. 172000025
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Dimethylglyoxime, 99+%, C4H8N2O2, CAS Number-95-45-4, 2,3-butanedione dioxime, chugaev's reagent, biacetyl, dioxime, 2,3-diisonitrosobutane, diacetyldioxime, nsc 9, diacetyl dioxime, unii-2971mft1ky, biacetyl dioxime, dimethylglyoxime, 500g, 01,772, White, 202-420-1, 116.12, GHS Signal Word: Danger | CAS: 95-45-4 | C4H8N2O2 | 116.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-45-4
Molecular Formula C4H8N2O2
Molecular Weight (g/mol) 116.12
MDL Number MFCD00002117
InChI Key JGUQDUKBUKFFRO-GGWOSOGESA-N
Synonym biacetyl dioxime, dimethylglyoxime, diacetyldioxime, 2,3-butanedione dioxime, diacetyl dioxime, 2,3-diisonitrosobutane, biacetyl, dioxime, chugaev's reagent, nsc 9, unii-2971mft1ky
PubChem CID 5356010
SMILES C\C(=N/O)\C(\C)=N\O

Specifications

Melting Point 239.0°C to 242.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic bottle
Linear Formula CH3C(=NOH)C(=NOH)CH3
Quantity 2.5 kg
Beilstein 01,772
Merck Index 15,327
Solubility Information Solubility in water: 0,6g/L 20°C. Other solubilities: soluble in alcohol,ether,pyridine and acetone,slightly soluble in benzene
Sublimation Point >70 °C
IUPAC Name N-[(E)-3-nitrosobut-2-en-2-yl]hydroxylamine
Formula Weight 116.12
Percent Purity 99+%
Insoluble Matter 0.05% max. (in alcohol)
Physical Form Crystalline Powder
Chemical Name or Material Dimethylglyoxime, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.