missing translation for 'onlineSavingsMsg'
Learn More

Dimethylglyoxime disodium salt octahydrate, Honeywell Fluka™

Product Code. 15625430 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
Product Code. Quantity unitSize
15625430 100 g 100g
15635430 500 g 500g
2 options
This item is not returnable. View return policy

Product Code. 15625430

Brand: Honeywell-Fluka 40400100G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

for precipitation of metals,≥97.0% (NT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75006-64-3
Molecular Formula C4H22N2Na2O10
Molecular Weight (g/mol) 304.20
MDL Number MFCD00150288
InChI Key IJJDRGUPRATWQX-UHFFFAOYSA-L
Synonym dimethylglyoxime disodium salt octahydrate, c4h6n2o2.2na.8h2o, sodium dimethylglyoxime octahydrate, dimethylglyoxime di-sodium salt octahydrate
PubChem CID 71310106
IUPAC Name disodium;2-N,3-N-dioxidobutane-2,3-diimine;octahydrate
SMILES O.O.O.O.O.O.O.O.[Na+].[Na+].CC(=N[O-])C(C)=N[O-]

Specifications

Quantity 100 g
Linear Formula CH3C(=NONa)C(=NONa)CH3 · 8H2O
UN Number NONH for all modes of transport
Beilstein 3776330
Formula Weight 304.2g/mol
Percent Purity ≥97% (NT)
Chemical Name or Material Dimethylglyoxime disodium salt octahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.