missing translation for 'onlineSavingsMsg'
Learn More

Dimethylphenylsilanol sodium salt, 97%

Product Code. 11472050
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11472050 5 g 5g
11462050 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11472050 Supplier Thermo Scientific Alfa Aesar Supplier No. H28457.06
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7646-75-5 | C8H11NaOSi | 174.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7646-75-5
Molecular Formula C8H11NaOSi
Molecular Weight (g/mol) 174.25
MDL Number MFCD09266151
InChI Key NPZJKOMUCXNLBN-UHFFFAOYSA-N
Synonym dimethylphenylsilanol sodium salt, sodium dimethyl phenyl silanolate, sodium dimethylphenylsilanolate, amtsi046, sodium phenyldimethylsilanolate, dimethyl phenyl sodiooxy silane, sodium phenyl dimethylsilanolate
PubChem CID 23713317
IUPAC Name sodium;dimethyl-oxido-phenylsilane
SMILES C[Si](C)(C1=CC=CC=C1)[O-].[Na+]

Specifications

Quantity 5 g
Formula Weight 174.25
Percent Purity 97%
Chemical Name or Material Dimethylphenylsilanol sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.