missing translation for 'onlineSavingsMsg'
Learn More

Dimethylsulfamoyl Chloride, 99%

Product Code. 10147470 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10147470 25 g Glass bottle 25g
10793114 100 g Glass bottle 100g
10793884 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10147470 Supplier Thermo Scientific Acros Supplier No. 408590250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13360-57-1 | C2H6ClNO2S | 143.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13360-57-1
Molecular Formula C2H6ClNO2S
Molecular Weight (g/mol) 143.59
MDL Number MFCD00007425
InChI Key JFCHSQDLLFJHOA-UHFFFAOYSA-N
Synonym dimethylsulfamoyl chloride, sulfamoyl chloride, dimethyl, dimethylaminosulfonyl chloride, dimethylsulfamoylchloride, dimethylsulphamoyl chloride, dimethylamidosulfonyl chloride, n,n-dimethylsulfamyl chloride, dimethylsulfamyl chloride, sulfamoyl chloride, n,n-dimethyl, dimethyl sulfamoyl chloride
PubChem CID 83372
IUPAC Name N,N-dimethylsulfamoyl chloride
SMILES CN(C)S(Cl)(=O)=O

Specifications

Melting Point -13°C
Density 1.3370g/mL
Boiling Point 114°C (75.0 mmHg)
Flash Point 94°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2NSO2Cl
Refractive Index 1.4510 to 1.453
Quantity 25 g
Beilstein 04, 84
Fieser 09,187
Specific Gravity 1.337
Solubility Information Solubility in water: decomposes in water
Formula Weight 143.59
Percent Purity 99%
Chemical Name or Material Dimethylsulfamoyl chloride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dimethylsulfamoyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H350-May cause cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Dimethylsulfamoyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.