missing translation for 'onlineSavingsMsg'
Learn More

Dimethylsulfamoyl Chloride, 99%

Product Code. 10793884 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10793884 500 g Glass bottle 500g
10147470 25 g Glass bottle 25g
10793114 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10793884 Supplier Thermo Scientific Acros Supplier No. 408595000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13360-57-1 | C2H6ClNO2S | 143.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13360-57-1
Molecular Formula C2H6ClNO2S
Molecular Weight (g/mol) 143.59
MDL Number MFCD00007425
InChI Key JFCHSQDLLFJHOA-UHFFFAOYSA-N
Synonym dimethylsulfamoyl chloride, sulfamoyl chloride, dimethyl, dimethylaminosulfonyl chloride, dimethylsulfamoylchloride, dimethylsulphamoyl chloride, dimethylamidosulfonyl chloride, n,n-dimethylsulfamyl chloride, dimethylsulfamyl chloride, sulfamoyl chloride, n,n-dimethyl, dimethyl sulfamoyl chloride
PubChem CID 83372
IUPAC Name N,N-dimethylsulfamoyl chloride
SMILES CN(C)S(Cl)(=O)=O

Specifications

Melting Point -13°C
Density 1.3370g/mL
Boiling Point 114°C (75.0 mmHg)
Flash Point 94°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2NSO2Cl
Refractive Index 1.4510 to 1.453
Quantity 500 g
Beilstein 04, 84
Fieser 09,187
Specific Gravity 1.337
Solubility Information Solubility in water: decomposes in water
Formula Weight 143.59
Percent Purity 99%
Chemical Name or Material Dimethylsulfamoyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.