missing translation for 'onlineSavingsMsg'
Learn More

Dimethylthiocarbamoyl chloride, 95%

Product Code. 15456578
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15456578 25 g 25g
15466578 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15456578 Supplier Thermo Scientific Alfa Aesar Supplier No. H60104.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 16420-13-6 | C3H6ClNS | 123.60 g/mol

Dimethylthiocarbamoyl chloride is used as an intermediate in the synthesis of pharmaceuticals. It finds application in chemoselective deoxygenation of pyridine-N-oxides. It is also used as a starting reagent in the synthesis of dimethylthiocarbamates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethylthiocarbamoyl chloride is used as an intermediate in the synthesis of pharmaceuticals. It finds application in chemoselective deoxygenation of pyridine-N-oxides. It is also used as a starting reagent in the synthesis of dimethylthiocarbamates.

Solubility
Soluble in chloroform and tetrahydrofuran.

Notes
Store in cool place. Moisture sensitive. Incompatible with strong oxidizing agents, amines and strong bases. Reacts violently with water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16420-13-6
Molecular Formula C3H6ClNS
Molecular Weight (g/mol) 123.60
MDL Number MFCD00004919
InChI Key PHWISQNXPLXQRU-UHFFFAOYSA-N
Synonym dimethylthiocarbamoyl chloride, carbamothioic chloride, dimethyl, chloromethanethioyl dimethylamine, n,n-dimethylthiocarbamoyl chloride, n,n-dimethylcarbamothioic chloride, dimethylthiocarbamoylchloride, carbamoyl chloride, dimethylthio, carbamothioic chloride, n,n-dimethyl, dimethylthiocarbamyl chloride, dimethylthiocarbamic acid chloride
PubChem CID 27871
SMILES CN(C)C(Cl)=S

Specifications

Melting Point 39°C to 43°C
Boiling Point 90°C to 95°C
Flash Point 98°C (208°F)
Linear Formula (CH3)2NCSCl
Quantity 25 g
UN Number UN3261
Beilstein 635823
Sensitivity Moisture sensitive
Solubility Information Soluble in chloroform and tetrahydrofuran.
IUPAC Name (chloromethanethioyl)dimethylamine
Formula Weight 123.6
Percent Purity 95%
Chemical Name or Material Dimethylthiocarbamoyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.