missing translation for 'onlineSavingsMsg'
Learn More

Dimethyltin Chloride, TRC

Product Code. 30370928
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30370928 1 g
30387036 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30370928 Supplier Toronto Research Chemicals Supplier No. TRCD4800301G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 753-73-1
Molecular Formula C2 H6 Cl2 Sn
Molecular Weight (g/mol) 219.68
Synonym Dimethyltin Dichloride, Stannane, dichlorodimethyl-, Tin, dimethyl-, dichloride (6CI), Dichlorodimethylstannane, Cotin 210, Dichlorodimethyltin, Dimethyldichlorostannane, Dimethyldichlorotin, Dimethylstannyl dichloride, Dimethyltin dichloride, Dimethyltin(IV) dichloride, NSC 55159, Nesstin P
IUPAC Name dichloro(dimethyl)stannane
SMILES C[Sn](C)(Cl)Cl

Specifications

Quantity 1 g
InChI Formula InChI=1S/2CH3.2ClH.Sn/h2*1H3;2*1H;/q;;;;+2/p-2
Formula Weight 219.8869
Chemical Name or Material Dimethyltin-dichloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.