missing translation for 'onlineSavingsMsg'
Learn More

Dimethyltin dichloride, 98%

Product Code. 11443238
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11443238 5 g 5g
11453238 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11443238 Supplier Thermo Scientific Alfa Aesar Supplier No. A17661.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 753-73-1 | C2H6Cl2Sn | 219.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 753-73-1
Molecular Formula C2H6Cl2Sn
Molecular Weight (g/mol) 219.68
MDL Number MFCD00000517
InChI Key PKKGKUDPKRTKLJ-UHFFFAOYSA-L
Synonym dimethyltin dichloride, dimethyldichlorotin, stannane, dichlorodimethyl, dichlorodimethyltin, dimethyltindichloride, dimethyldichlorostannane, dichloro dimethyl stannane, tin, dimethyl-, dichloride, cotin 210, dichlorid dimethylcinicity
PubChem CID 12955
IUPAC Name dichloro(dimethyl)stannane
SMILES C[Sn](C)(Cl)Cl

Specifications

Melting Point 104°C to 108°C
Boiling Point 188°C to 190°C
Quantity 5 g
UN Number UN3146
Beilstein 3660508
Sensitivity Moisture sensitive
Formula Weight 219.69
Percent Purity 98%
Chemical Name or Material Dimethyltin dichloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.