missing translation for 'onlineSavingsMsg'
Learn More

Dimidium bromide, 95%

Product Code. 11406586
Change view
Click to view available options
Quantity:
0.25 g
1 g
Unit Size:
0.25g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11406586 0.25 g 0.25g
11496576 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11406586 Supplier Thermo Scientific Alfa Aesar Supplier No. B24818.01
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 518-67-2 | C20H18BrN3 | 380.29 g/mol

Dimidium bromide acts as an inhibitor of L-cell growth. It is also used as a fluorescent probe for nucleic acids. It is also useful in the determination of cationic surfactants. It is also used as an intercalating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimidium bromide acts as an inhibitor of L-cell growth. It is also used as a fluorescent probe for nucleic acids. It is also useful in the determination of cationic surfactants. It is also used as an intercalating agent.

Solubility
Soluble in water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 518-67-2
Molecular Formula C20H18BrN3
Molecular Weight (g/mol) 380.29
MDL Number MFCD00011757
InChI Key MQOKYEROIFEEBH-UHFFFAOYSA-N
Synonym dimidium bromide, trypadine, 3,8-diamino-5-methyl-6-phenylphenanthridinium bromide, phenanthridinium 1553, phenanthridinium compound 1553, phenanthridinium, 3,8-diamino-5-methyl-6-phenyl-, bromide, 2,7-diamino-10-methyl-9-phenylphenanthridinium bromide, 3,8-diamino-5-methyl-6-phenylphenanthridin-5-ium bromide, phenanthridium 1553
PubChem CID 68207
IUPAC Name 5-methyl-6-phenylphenanthridin-5-ium-3,8-diamine;bromide
SMILES [Br-].C[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12

Specifications

Melting Point 230°C (decomposition)
Quantity 0.25 g
Formula Weight 380.31
Percent Purity 95%
Chemical Name or Material Dimidium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.