missing translation for 'onlineSavingsMsg'
Learn More

Dimidium bromide, 98%

Product Code. 10753724 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10753724 1 g Glass bottle 1g
10174790 250 mg Glass bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10753724 Supplier Thermo Scientific Acros Supplier No. 215340010
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 518-67-2 | C20H18BrN3 | 380.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 518-67-2
Molecular Formula C20H18BrN3
Molecular Weight (g/mol) 380.29
MDL Number MFCD00011757
InChI Key MQOKYEROIFEEBH-UHFFFAOYSA-N
Synonym dimidium bromide, trypadine, 3,8-diamino-5-methyl-6-phenylphenanthridinium bromide, phenanthridinium 1553, phenanthridinium compound 1553, phenanthridinium, 3,8-diamino-5-methyl-6-phenyl-, bromide, 2,7-diamino-10-methyl-9-phenylphenanthridinium bromide, 3,8-diamino-5-methyl-6-phenylphenanthridin-5-ium bromide, phenanthridium 1553
PubChem CID 68207
IUPAC Name 5-methyl-6-phenylphenanthridin-5-ium-3,8-diamine;bromide
SMILES [Br-].C[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12

Specifications

Melting Point 230°C
Color Purple to Red
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (on dry substance) (Argentometry)
Packaging Glass bottle
Quantity 1 g
Beilstein 22, IV, 5515
Merck Index 09, 3257
Solubility Information Solubility in water: soluble
Formula Weight 380.28
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Dimidium bromide, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Dimidium bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 3,8-Diamino-5-methyl-6-phenylphenanthridinium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.