Learn More
Lenz Laborglasintrumente Dimroth Condensers for Extractors, with Olives
Product Code. 10393191
Change view
Click to view available options
Standard Taper Joint No:
34/35 NS
71/51 NS
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No Unit Size
10393191 34/35 NS Each
10261981 71/51 NS Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10393191 Supplier Lenz Laborglasintrumente Supplier No. 05246005
  • ONLINE EXCLUSIVE PRICE
    152.00 €
    95.97 € / Pack of 1
    Save 56.03 €
    36% Off
Estimated Shipment: 18-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Dimroth condensers for extractors
  • For condensing vapours
  • With NS cone
  • Water connection: glass olives
  • Made of DURAN™ tubing
Specifications
Includes Glass olives
Quantity 1 Ea.
Standard Taper Joint No 34/35 NS
Material Duran Glass