Learn More
Lenz Laborglasintrumente Dimroth Condensers, with GL Threads
Product Code. 11506814
Change view
Click to view available options
Standard Taper Joint No:
NS 14/23
NS 24/29
NS 45/40
Length (Metric):
250 mm
400 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No Length (Metric) unitSize
11506814 NS 14/23 250 mm Each
10528954 NS 24/29 250 mm Each
11526814 NS 45/40 250 mm Each
11536814 NS 45/40 400 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11506814 Supplier Lenz Laborglasintrumente Supplier No. 05240311
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Dimroth condensers acc. to DIN 12591
  • For condensing vapours
  • With NS socket and NS cone
  • Water connection: GL 14 thread (with two threaded plastic olives)
  • Made of DURAN™ tubing
Specifications
Length (Metric) 250 mm
Standard Taper Joint No NS 14/23
Material Duran Glass