missing translation for 'onlineSavingsMsg'
Learn More

Dioctyloxotin >90%, TRC

Product Code. 30385911
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30385911 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385911 Supplier Toronto Research Chemicals Supplier No. TRCD68156510G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dioctyloxotin
CAS 870-08-6
Molecular Formula C16H34OSn
Quantity 10 g
Synonym Irgastab T 161, NSC 140743, Neostann U 800P, Stann OO, U 800, U 800 (heat stabilizer), Di-n-octyltin oxidel Dioctyloxostannane, DOTO (dioctyltin oxide), DOTO
InChI Formula InChI=1S/2C8H17.O.Sn/c2*1-3-5-7-8-6-4-2;;/h2*1,3-8H2,2H3;;
SMILES CCCCCCCC[Sn](=O)CCCCCCCC
IUPAC Name dioctyl(oxo)tin
Molecular Weight (g/mol) 361.15
Formula Weight 362.1632
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.