Learn More
Thermo Scientific™ Dionex™ OnGuard™ II H Cartridges
Product Code. 11331753
Change view
Click to view available options
Quantity:
1
Unit Size:
Pack of 48
Product Code. Quantity Unit Size
11331753 1 Pack of 48
1 options
Product Code. 11331753 Supplier Thermo Scientific™ Supplier No. 057086
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Remove alkaline earth, transition metals and neutralize highly alkaline samples to improve IC performance and facilitate trace analyses. Pretreat up to 12 samples in parallel.
Remove high levels of alkaline earth and transition metals from sample matrices and neutralize highly alkaline samples such as sodium hydroxide or sodium carbonate. Carbonate can then be removed by sparging the sample. The Thermo Scientific™ Dionex™ OnGuard™ II H Cartridge is an off-line sample preparation technique that eliminates these matrix interferences to improve ion chromatography (IC) performance and facilitate low-level ion analysis. Use them singly or in series, or pretreat up to 12 samples in parallel using the optional Thermo Scientific Dionex OnGuard Sample Prep Station.
Cartridges use a 16% cross-linked, styrene-based, sulfonic acid resin in the hydrogen form with high selectivity for multivalent cations such as calcium and transition metals.
Solve Tough Application Problems
  • Boost sensitivity, lower baselines, and get better separations by removing phenolics, metals, cations, anions, hydrophobic substances, and more
  • Achieve reproducible trace-level determinations in concentrated matrices
  • Increase the lifetime of your analytical columns
  • Use for a wide range of matrices thanks to wide pH stability
  • Supports applications with choice of 1cc and 2.5cc formats, as well as syringe barrel format for robotic processing (custom order)
Easy-to-Use, Leak-Free
  • Leak-free, no-channeling cartridge
  • Quick, secure Luer inlet connections
Performance Guaranteed
  • Ships with a Certificate of Analysis that verifies capacity, performance and cleanliness
Pretreat 12 Samples Simultaneously
  • Use the optional Dionex OnGuard Sample Prep Station and OnGuard Sample Pretreatment Cartridges
Specifications
Description H; 2.5 cc
For Use With (Application) Removal of High Levels of Alkaline Earth and Transition Metals from Sample Matrices
For Use With (Equipment) Dionex OnGuard Sample Prep Station
Material Cross-Linked, Styrene-Based, Sulfonic Acid Resin in the Hydrogen Form
Volume (Metric) 2.5 mL
Quantity 1
Product Line OnGuard