Learn More
Thermo Scientific™ Dionex™ Specially Formulated PTFE Tubing
Product Code. 11322133 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
250 and 350 um ID
250 um ID; Low pressure
350 um ID; Low pressure
For Use With (Equipment):
Dionex UltiMate 3000 RSLC Nano System
Fused Silica Capillary Tubing
Unit Size:
Pack of 2
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description For Use With (Equipment) Unit Size
11322133 250 um ID; Low pressure Dionex UltiMate 3000 RSLC Nano System Pack of 5
11352133 250 and 350 um ID Fused Silica Capillary Tubing Pack of 2
11362143 350 um ID; Low pressure Fused Silica Capillary Tubing Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11322133 Supplier Thermo Scientific™ Supplier No. 160486
Estimated Shipment: 23-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Thermo Scientific™ Dionex Specially Formulated PTFE Tubing is used as connector for fused silica tubing (nano and cap LC). Choose fitting dimensions for OD of fused silica tubing used.
Thermo Scientific™ Dionex Specially Formulated PTFE Tubing is used as connector for fused silica tubing (nano and cap LC).
TRUSTED_SUSTAINABILITY
Specifications
Description 250 um ID; Low pressure
For Use With (Equipment) Dionex UltiMate 3000 RSLC Nano System
Product Line nanoLC