missing translation for 'onlineSavingsMsg'
Learn More

Dioscin, MedChemExpress

Product Code. 16404767
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16404767 50 mg 50mg
16498868 10 mg 10mg
16491308 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16404767 Supplier Medchem Express Supplier No. HYN0124

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dioscin(CCRIS 4123; Collettiside III) is a natural steroid saponin derived from several plants, showing potent anti-cancer effect against a variety of tumor cell lines.

Dioscin(CCRIS 4123

Specifications

Chemical Name or Material Dioscin
CAS 19057-60-4
Color White
Purity Grade Notes Research
Molecular Formula C45H72O16
Quantity 50 mg
Synonym Collettiside III CCRIS 4123
Solubility Information DMSO : ≥ 100 mg/mL (115.07 mM)
SMILES O[C@@H]1[C@@H](O)[C@H](C)O[C@@]([H])(O[C@H]2[C@H](O)[C@@H](O[C@]3([H])O[C@@H](C)[C@H](O)[C@@H](O)[C@H]3O)[C@@](O[C@@H]4CC5=CC[C@]([C@]([H])(C[C@@]6([H])[C@]7([H])[C@H](C)[C@]8(OC[C@H](C)CC8)O6)[C@]7(C)CC9)([H])[C@@]9([H])[C@@]5(C)CC4)([H])O[C@@H]2CO)[C@@H]1O
Molecular Weight (g/mol) 869.04
Formula Weight 869.04
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.