Learn More
Diosmin, Thermo Scientific™
49.90 € - 130.00 €
Chemical Identifiers
| CAS | 520-27-4 |
|---|---|
| Molecular Formula | C28H32O15 |
| Molecular Weight (g/mol) | 608.549 |
| MDL Number | MFCD00009772 |
| InChI Key | GZSOSUNBTXMUFQ-YFAPSIMESA-N |
| Synonym | diosmin, barosmin, diosmine, venosmine, diosmil, flebosten, daflon, diosimin, diosminum, tovene |
| PubChem CID | 5281613 |
| ChEBI | CHEBI:4631 |
| IUPAC Name | 5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one |
| SMILES | CC1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3=CC(=C4C(=C3)OC(=CC4=O)C5=CC(=C(C=C5)OC)O)O)O)O)O)O)O)O |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15484519
|
Thermo Scientific Alfa Aesar
J62073.06 |
5g |
49.90 €
5g |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
15494519
|
Thermo Scientific Alfa Aesar
J62073.14 |
25g |
130.00 €
25g |
Estimated Shipment: 10-09-2026
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
SolubilitySoluble in DMSO (50 mg/ml), water (122 mg/ml at 25°C), and ethanol (<1mg/mL at 25°C).
Notes
Protect from heat. Incompatible with heat and oxidizing agents. Store at 4°C.
Chemical Identifiers
| 520-27-4 | |
| 608.549 | |
| GZSOSUNBTXMUFQ-YFAPSIMESA-N | |
| 5281613 | |
| 5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one |
| C28H32O15 | |
| MFCD00009772 | |
| diosmin, barosmin, diosmine, venosmine, diosmil, flebosten, daflon, diosimin, diosminum, tovene | |
| CHEBI:4631 | |
| CC1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3=CC(=C4C(=C3)OC(=CC4=O)C5=CC(=C(C=C5)OC)O)O)O)O)O)O)O)O |
Specifications
| 520-27-4 | |
| MFCD00009772 | |
| diosmin, barosmin, diosmine, venosmine, diosmil, flebosten, daflon, diosimin, diosminum, tovene | |
| GZSOSUNBTXMUFQ-YFAPSIMESA-N | |
| 5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one | |
| 5281613 | |
| 608.5 |
| C28H32O15 | |
| 14,3297 | |
| Soluble in DMSO (50mg/ml),water (122mg/ml at 25°C),and ethanol (<1mg/ml at 25°C). | |
| CC1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3=CC(=C4C(=C3)OC(=CC4=O)C5=CC(=C(C=C5)OC)O)O)O)O)O)O)O)O | |
| 608.549 | |
| CHEBI:4631 | |
| Diosmin |
Safety and Handling
missing translation for 'einecsNumber' : 208-289-7
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.