missing translation for 'onlineSavingsMsg'
Learn More

Diosmin, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar J62073.14

Quantity 25g

 View more versions of this product

Product Code. 15494519

  • 130.00 € / 25g
 Restricted Item
Estimated Shipment: 10-09-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Soluble in DMSO (50 mg/ml), water (122 mg/ml at 25°C), and ethanol (<1mg/mL at 25°C).

Notes
Protect from heat. Incompatible with heat and oxidizing agents. Store at 4°C.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

520-27-4
608.549
GZSOSUNBTXMUFQ-YFAPSIMESA-N
5281613
5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one
C28H32O15
MFCD00009772
diosmin, barosmin, diosmine, venosmine, diosmil, flebosten, daflon, diosimin, diosminum, tovene
CHEBI:4631
CC1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3=CC(=C4C(=C3)OC(=CC4=O)C5=CC(=C(C=C5)OC)O)O)O)O)O)O)O)O

Specifications

520-27-4
MFCD00009772
14,3297
Soluble in DMSO (50mg/ml),water (122mg/ml at 25°C),and ethanol (<1mg/ml at 25°C).
CC1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3=CC(=C4C(=C3)OC(=CC4=O)C5=CC(=C(C=C5)OC)O)O)O)O)O)O)O)O
608.549
CHEBI:4631
Diosmin
C28H32O15
25g
diosmin, barosmin, diosmine, venosmine, diosmil, flebosten, daflon, diosimin, diosminum, tovene
GZSOSUNBTXMUFQ-YFAPSIMESA-N
5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one
5281613
608.5
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.