missing translation for 'onlineSavingsMsg'
Learn More

Dioxindolyl-L-alanine (Mixture of Diastereomers), TRC

Product Code. 30006820
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30006820 500 mg 500mg
30005620 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30006820 Supplier Toronto Research Chemicals Supplier No. TRCD486855500MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 184955-21-3
Molecular Formula C11 H12 N2 O4
Molecular Weight (g/mol) 236.22
Synonym (αS)-α-Amino-2,3-dihydro-3-hydroxy-2-oxo-1H-indole-3-propanoic Acid, [3(S)]-α-Amino-2,3-dihydro-3-hydroxy-2-oxo-1H-indole-3-propanoic Acid;
IUPAC Name (2S)-2-amino-3-(3-hydroxy-2-oxo-1H-indol-3-yl)propanoic acid
SMILES N[C@@H](CC1(O)C(=O)Nc2ccccc12)C(=O)O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C11H12N2O4/c12-7(9(14)15)5-11(17)6-3-1-2-4-8(6)13-10(11)16/h1-4,7,17H,5,12H2,(H,13,16)(H,14,15)/t7-,11?/m0/s1
Formula Weight 236.0797
Percent Purity >95%
Chemical Name or Material (2S)-2-amino-3-(3-hydroxy-2-oxo-1H-indol-3-yl)propanoic acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.