missing translation for 'onlineSavingsMsg'
Learn More

Dipentene, tech., Thermo Scientific Chemicals

Product Code. 15422457
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15422457 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15422457 Supplier Thermo Scientific Alfa Aesar Supplier No. 045805.AK
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dipentene is a monoterpene olefin having potential applications in polymer and fuel chemistry. It is also widely used as flavoring and fragrance agent. It is a promising green solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138-86-3
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00062992
InChI Key XMGQYMWWDOXHJM-UHFFFAOYNA-N
Synonym dipentene, limonene, cinene, cajeputene, dl-limonene, kautschin, p-mentha-1,8-diene, dipenten, polylimonene, eulimen
PubChem CID 22311
ChEBI CHEBI:15384
SMILES CC(=C)C1CCC(C)=CC1

Specifications

Density 0.85 g/mL
Boiling Point 170°C to 180°C
Flash Point 43°C (109°F)
Linear Formula CH3C6H8C(CH3)=CH2
Refractive Index 1.474
Quantity 250 mL
UN Number UN2052
Beilstein 3587825
Merck Index 14,5493
Solubility Information Soluble in ether (miscible) and ethanol (miscible). Insoluble in water.
IUPAC Name 1-methyl-4-prop-1-en-2-ylcyclohexene
Formula Weight 136.24
Grade Technical
Physical Form Liquid
Chemical Name or Material Dipentene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.