missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl Disulfide, TRC

Product Code. 30134322
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134322 5 g 5g
17921895 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134322 Supplier Toronto Research Chemicals Supplier No. TRCD4876305G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 882-33-7
Molecular Formula C12 H10 S2
Molecular Weight (g/mol) 218.34
Synonym Phenyl disulfide (8CI), Diphenyl disulfide, (Phenyldisulfanyl)benzene, 1,1'-Dithiodibenzene, 1,2-Diphenyldisulfane, Biphenyl disulfide, DPDS, Diphenyl disulphide, NSC 2689, Diphenyl Disulfide
IUPAC Name (phenyldisulfanyl)benzene
SMILES S(Sc1ccccc1)c2ccccc2

Specifications

Quantity 5 g
InChI Formula InChI=1S/C12H10S2/c1-3-7-11(8-4-1)13-14-12-9-5-2-6-10-12/h1-10H
Formula Weight 218.0224
Percent Purity >95%
Chemical Name or Material Diphenyl disulfide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.