missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl phosphate, 99%

Product Code. 10607552 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10607552 5 g Glass bottle 5g
10328410 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10607552 Supplier Thermo Scientific Acros Supplier No. 225690050
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 838-85-7 | C12H11O4P | 250.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 838-85-7
Molecular Formula C12H11O4P
Molecular Weight (g/mol) 250.19
MDL Number MFCD00003033
InChI Key ASMQGLCHMVWBQR-UHFFFAOYSA-N
Synonym diphenyl phosphate, phosphoric acid, diphenyl ester, phenyl hydrogen phosphate, phenyl phosphate pho 2 ho po, diphenoxyphosphinic acid, phosphoric acid diphenyl ester, diphenylphosphoric acid, phosphoric acid diphenyl, dsstox_cid_28182
PubChem CID 13282
IUPAC Name diphenyl hydrogen phosphate
SMILES OP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1

Specifications

Melting Point 63°C to 70°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (C6H5O)2P(O)OH
Quantity 5 g
Beilstein 06, 178
Formula Weight 250.19
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Diphenyl phosphate, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.