missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl Phthalate, TRC

Product Code. 30377012
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30377012 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30377012 Supplier Toronto Research Chemicals Supplier No. TRCD4915905G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phthalic acid, bis-phenyl ester
CAS 84-62-8
Molecular Formula C20 H14 O4
Quantity 5 g
Synonym 1,2-Benzenedicarboxylic acid, 1,2-diphenyl ester, 1,2-Benzenedicarboxylic acid, diphenyl ester (9CI), Phthalic acid, diphenyl ester (6CI,7CI,8CI), 1,2-Diphenyl benzene-1,2-dicarboxylate, Diphenyl phthalate, NSC 4052, Phenyl phthalate, Phthalic acid, bis-phenyl ester, Phthalic acid diphenyl ester
InChI Formula InChI=1S/C20H14O4/c21-19(23-15-9-3-1-4-10-15)17-13-7-8-14-18(17)20(22)24-16-11-5-2-6-12-16/h1-14H
SMILES O=C(Oc1ccccc1)c2ccccc2C(=O)Oc3ccccc3
IUPAC Name diphenyl benzene-1,2-dicarboxylate
Molecular Weight (g/mol) 318.32
Formula Weight 318.0892
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.