missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl sulfide, 98%

Product Code. 11419003
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419003 25 g 25g
11429003 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11419003 Supplier Thermo Scientific Alfa Aesar Supplier No. A11114.14
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 139-66-2 | C12H10S | 186.27 g/mol

Diphenyl sulfide is used as a raw material for triaryl sulfonium photoinitiator, intermediates for agrichemicals, catalyst, metal extractants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenyl sulfide is used as a raw material for triaryl sulfonium photoinitiator, intermediates for agrichemicals, catalyst, metal extractants.

Solubility
Soluble in ether, benzene, carbon disulphide.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents, acids, acid chlorides, acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139-66-2
Molecular Formula C12H10S
Molecular Weight (g/mol) 186.27
MDL Number MFCD00003064
InChI Key LTYMSROWYAPPGB-UHFFFAOYSA-N
Synonym diphenyl sulfide, phenyl sulfide, phenylthiobenzene, diphenyl sulphide, diphenyl thioether, benzene, 1,1'-thiobis, diphenylmercaptan, diphenylthiamethane, diphenyl monosulfide, sulfide, diphenyl
PubChem CID 8766
ChEBI CHEBI:38959
IUPAC Name phenylsulfanylbenzene
SMILES S(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point -40°C
Density 1.118
Boiling Point 295°C to 296°C
Flash Point >110°C (230°F)
Odor Unpleasant
Refractive Index 1.6325
Quantity 25 g
Beilstein 1907932
Merck Index 14,7314
Solubility Information Soluble in ether,benzene,carbon disulphide.
Formula Weight 186.28
Percent Purity 98%
Chemical Name or Material Diphenyl sulfide
Show More Show Less
compliance-icons
Product Identifier
  • Diphenyl sulfide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Phenyl sulfide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.