missing translation for 'onlineSavingsMsg'
Learn More

Diphenylacetylene, 99%

Product Code. 10703851 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10703851 5 g Glass bottle 5g
10764221 25 g Glass bottle 25g
11974451 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10703851 Supplier Thermo Scientific Acros Supplier No. 117180050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 501-65-5 | C14H10 | 178.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 501-65-5
Molecular Formula C14H10
Molecular Weight (g/mol) 178.23
InChI Key JRXXLCKWQFKACW-UHFFFAOYSA-N
Synonym diphenylacetylene, tolan, diphenylethyne, 1,2-diphenylethyne, tolane, 1,2-diphenylacetylene, benzene, 1,1'-1,2-ethynediyl bis, biphenylacetylene, diphenyl acetylene, ethyne, diphenyl
PubChem CID 10390
ChEBI CHEBI:51579
IUPAC Name 2-phenylethynylbenzene
SMILES C1=CC=C(C=C1)C#CC2=CC=CC=C2

Specifications

Melting Point 58.0°C to 61.0°C
Color Brown to Yellow
Density 0.9900g/mL
Boiling Point 170.0°C (19.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5C≡CC6H5
Quantity 5 g
Beilstein 05,656
Fieser 01,335
Merck Index 15,9664
Specific Gravity 0.99
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether and hot alcohol
Formula Weight 178.23
Percent Purity 99%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material Diphenylacetylene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.