missing translation for 'onlineSavingsMsg'
Learn More

Diphenylacetylene, TRC

Product Code. 30372969
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30372969 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30372969 Supplier Toronto Research Chemicals Supplier No. TRCD4933985G
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diphenylacetylene
CAS 501-65-5
Molecular Formula C14H10
Quantity 5 g
Synonym 1,2-Diphenylacetylene, 1,2-Diphenylethyne, Diphenylacetylene, Diphenylethyne, Diphenylethyne, NSC 5185, Tolan, Tolane
InChI Formula InChI=1S/C14H10/c1-3-7-13(8-4-1)11-12-14-9-5-2-6-10-14/h1-10H
SMILES c1ccc(cc1)C#Cc2ccccc2
Molecular Weight (g/mol) 178.23
Formula Weight 178.0783
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.